Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrd1
Entrez Gene ID
100758524
Gene Name
leucine-rich repeats and death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613638: 1466889 - 1487716 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496946.2 XP_003496994.1 2 - 3 1944 71

View Full Product Details