Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dkk1
Entrez Gene ID
100769380
Gene Name
dickkopf WNT signaling pathway inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613782: 1750895 - 1753046 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500227.1 XP_003500275.1 3 - 4 556 94

View Full Product Details