Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mon1b
Entrez Gene ID
100761265
Gene Name
MON1 secretory trafficking family member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613802: 1197015 - 1206650 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500660.2 XP_003500708.1 1 - 2 552 97
XM_007641413.1 XP_007639603.1 2 - 3 308 97
XM_007641414.1 XP_007639604.1 2 - 3 336 97

View Full Product Details