Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina4
Entrez Gene ID
100763387
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4
Gene Aliases
I79_007747
Chromosome Location
Chr.Un|NW_003613846: 23822 - 28601 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501366.1 XP_003501414.1 2 - 3 905 74

View Full Product Details