Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpr97
Entrez Gene ID
100764587
Gene Name
G protein-coupled receptor 97
Gene Aliases
I79_018013
Chromosome Location
Chr.Un|NW_003614770: 301597 - 327136 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509802.1 XP_003509850.1 11 - 12 1510 69

View Full Product Details