Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Them4
Entrez Gene ID
100769809
Gene Name
thioesterase superfamily member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615668: 168646 - 191522 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512953.1 XP_003513001.1 2 - 3 254 89

View Full Product Details