Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cebpe
Entrez Gene ID
100770450
Gene Name
CCAAT/enhancer binding protein (C/EBP), epsilon
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614213: 795253 - 797457 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506128.2 XP_003506176.1 1 - 2 1031 115

View Full Product Details