Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm4sf20
Entrez Gene ID
100772323
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617096: 74216 - 80916 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514904.1 XP_003514952.1 1 - 2 68 90

View Full Product Details