Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scn2b
Entrez Gene ID
100767440
Gene Name
sodium channel, voltage-gated, type II, beta subunit
Gene Aliases
I79_024769
Chromosome Location
Chr.Un|NW_003617676: 37677 - 45289 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515274.1 XP_003515322.1 1 - 2 65 79

View Full Product Details