Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr88
Entrez Gene ID
100768497
Gene Name
WD repeat domain 88
Gene Aliases
I79_025072
Chromosome Location
Chr.Un|NW_003618259: 10263 - 36834 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607670.1 XP_007605860.1 8 - 9 811 94

View Full Product Details