Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lypd1
Entrez Gene ID
100759295
Gene Name
LY6/PLAUR domain containing 1
Gene Aliases
I79_003213
Chromosome Location
Chr.Un|NW_003613656: 1350428 - 1388528 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497613.2 XP_003497661.1 2 - 3 188 71

View Full Product Details