Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aplnr
Entrez Gene ID
100759527
Gene Name
apelin receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614216: 840906 - 842039 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506166.1 XP_003506214.1 1 - 1 1018 64

View Full Product Details