Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tstd1
Entrez Gene ID
100764675
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614301: 105778 - 107278 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506852.2 XP_003506900.1 2 - 3 483 75

View Full Product Details