Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfrsf17
Entrez Gene ID
100753580
Gene Name
tumor necrosis factor receptor superfamily, member 17
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613631: 1278707 - 1296399 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496711.2 XP_003496759.2 1 - 2 146 82

View Full Product Details