Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scnm1
Entrez Gene ID
100769370
Gene Name
sodium channel modifier 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613699: 2275499 - 2279171 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498667.2 XP_003498715.1 4 - 5 422 101
GenBank mRNA JI863698.1 - 4 - 5 422 101
JP057693.1 - 4 - 5 422 101

View Full Product Details