Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc30
Entrez Gene ID
100762333
Gene Name
leucine rich repeat containing 30
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613991: 1313363 - 1314265 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503697.1 XP_003503745.1 1 - 1 725 85

View Full Product Details