Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adora2a
Entrez Gene ID
100762652
Gene Name
adenosine A2a receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614478: 33482 - 41816 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508085.2 XP_003508133.1 1 - 2 321 68

View Full Product Details