Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps37c
Entrez Gene ID
100763936
Gene Name
vacuolar protein sorting 37 homolog C (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003618059: 10468 - 37875 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515445.2 XP_003515493.1 2 - 3 113 79
XM_007607616.1 XP_007605806.1 3 - 4 316 79
XM_007607617.1 XP_007605807.1 3 - 4 291 79
XM_007607618.1 XP_007605808.1 2 - 3 142 79
XM_007607619.1 XP_007605809.1 2 - 3 113 79
XM_007607621.1 XP_007605811.1 2 - 3 114 79
GenBank mRNA JI863404.1 - 1 - 2 75 79
JP055181.1 - 1 - 2 75 79

View Full Product Details