Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dpep3
Entrez Gene ID
100756154
Gene Name
dipeptidase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614050: 1065092 - 1070030 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644849.1 XP_007643039.1 9 - 10 1218 60

View Full Product Details