Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc25
Entrez Gene ID
100768903
Gene Name
leucine rich repeat containing 25
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613716: 915654 - 916451 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499113.1 XP_003499161.1 1 - 1 722 87

View Full Product Details