Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chad
Entrez Gene ID
100758456
Gene Name
chondroadherin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613846: 1662572 - 1667390 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501352.2 XP_003501400.1 1 - 2 1638 78

View Full Product Details