Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk6
Entrez Gene ID
100764262
Gene Name
potassium channel, subfamily K, member 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614005: 388652 - 405609 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503830.1 XP_003503878.1 1 - 2 178 71

View Full Product Details