Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a13
Entrez Gene ID
100754527
Gene Name
solute carrier family 22 (organic anion/urate transporter), member 13
Gene Aliases
I79_013207
Chromosome Location
Chr.Un|NW_003614199: 107268 - 110914 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505936.1 XP_003505984.1 3 - 4 634 84

View Full Product Details