Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myl6b
Entrez Gene ID
100766814
Gene Name
myosin, light chain 6B, alkali, smooth muscle and non-muscle
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614217: 553495 - 558501 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506188.2 XP_003506236.1 3 - 4 327 74

View Full Product Details