Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppapdc3
Entrez Gene ID
100751688
Gene Name
phosphatidic acid phosphatase type 2 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615645: 199297 - 211440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512895.2 XP_003512943.1 1 - 2 696 59

View Full Product Details