Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Utp3
Entrez Gene ID
100774325
Gene Name
UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614817: 107018 - 108652 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510045.2 XP_003510093.1 1 - 1 307 68

View Full Product Details