Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnrh1
Entrez Gene ID
100774729
Gene Name
gonadotropin-releasing hormone 1 (luteinizing-releasing hormone)
Gene Aliases
I79_001531
Chromosome Location
Chr.Un|NW_003613613: 4234067 - 4238851 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496189.2 XP_003496237.2 2 - 3 1768 61

View Full Product Details