Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam53c
Entrez Gene ID
100773129
Gene Name
family with sequence similarity 53, member C
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613902: 645745 - 657406 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502518.2 XP_003502566.1 2 - 3 271 78

View Full Product Details