Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g2d
Entrez Gene ID
100754712
Gene Name
phospholipase A2, group IID
Gene Aliases
I79_014444
Chromosome Location
Chr.Un|NW_003614302: 749771 - 755611 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506898.1 XP_003506946.1 1 - 2 35 93

View Full Product Details