Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes6
Entrez Gene ID
100772285
Gene Name
hes family bHLH transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614689: 283327 - 284920 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509329.2 XP_003509377.1 1 - 2 191 64

View Full Product Details