Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgls
Entrez Gene ID
100774660
Gene Name
6-phosphogluconolactonase
Gene Aliases
I79_005134
Chromosome Location
Chr.Un|NW_003613716: 1323894 - 1326043 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007638583.1 XP_007636773.1 2 - 3 219 86

View Full Product Details