Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Maml3
Entrez Gene ID
100767462
Gene Name
mastermind-like 3 (Drosophila)
Gene Aliases
I79_007545
Chromosome Location
Chr.Un|NW_003613834: 1567412 - 1743700 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641861.1 XP_007640051.1 4 - 5 1902 142

View Full Product Details