Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo45
Entrez Gene ID
100761316
Gene Name
F-box protein 45
Gene Aliases
I79_000627
Chromosome Location
Chr.Un|NW_003613591: 1782813 - 1801643 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495427.2 XP_003495475.2 2 - 3 517 71

View Full Product Details