Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm4sf1
Entrez Gene ID
100758297
Gene Name
transmembrane 4 L six family member 1
Gene Aliases
I79_015994
Chromosome Location
Chr.Un|NW_003614490: 305647 - 313391 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508146.2 XP_003508194.1 3 - 4 708 78
GenBank mRNA JI879266.1 - 1 - 2 100 78

View Full Product Details