Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt9
Entrez Gene ID
100763915
Gene Name
synaptotagmin IX
Gene Aliases
I79_022480
Chromosome Location
Chr.Un|NW_003615961: 15780 - 120633 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513487.1 XP_003513535.1 5 - 6 1286 69

View Full Product Details