Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppap2b
Entrez Gene ID
100774527
Gene Name
phosphatidic acid phosphatase type 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613599: 1782139 - 1859969 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495751.2 XP_003495799.1 1 - 2 389 68
GenBank mRNA JI869948.1 - 1 - 2 536 68
JP063324.1 - 1 - 2 536 68

View Full Product Details