Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March4
Entrez Gene ID
100754006
Gene Name
membrane-associated ring finger (C3HC4) 4, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613713: 1069492 - 1179445 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498994.2 XP_003499042.1 1 - 2 506 57

View Full Product Details