Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prcd
Entrez Gene ID
100755980
Gene Name
progressive rod-cone degeneration
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614300: 734397 - 747194 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506826.2 XP_003506874.1 4 - 5 706 93

View Full Product Details