Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec1a
Entrez Gene ID
100772475
Gene Name
C-type lectin domain family 1, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614673: 188863 - 213389 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509256.2 XP_003509304.2 5 - 6 656 67

View Full Product Details