Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vcpip1
Entrez Gene ID
100763244
Gene Name
valosin containing protein (p97)/p47 complex interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615083: 105996 - 131724 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511341.2 XP_003511389.2 2 - 3 2227 91

View Full Product Details