Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klb
Entrez Gene ID
100758821
Gene Name
klotho beta
Gene Aliases
I79_024816
Chromosome Location
Chr.Un|NW_003617759: 38645 - 53395 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515313.2 XP_003515361.2 3 - 4 926 105

View Full Product Details