Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ipcef1
Entrez Gene ID
100774344
Gene Name
interaction protein for cytohesin exchange factors 1
Gene Aliases
I79_001180
Chromosome Location
Chr.Un|NW_003613604: 3029775 - 3085519 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495893.1 XP_003495941.1 7 - 8 991 74

View Full Product Details