Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnt16
Entrez Gene ID
100764115
Gene Name
wingless-type MMTV integration site family, member 16
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615657: 11690 - 21520 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512934.1 XP_003512982.1 1 - 2 92 82

View Full Product Details