Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm46
Entrez Gene ID
100769270
Gene Name
RNA binding motif protein 46
Gene Aliases
I79_005567
Chromosome Location
Chr.Un|NW_003613739: 994771 - 1031701 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499468.2 XP_003499516.1 1 - 2 154 104
XM_007640222.1 XP_007638412.1 1 - 2 154 104
XM_007640223.1 XP_007638413.1 1 - 2 154 104

View Full Product Details