Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Naglu
Entrez Gene ID
100757611
Gene Name
N-acetylglucosaminidase, alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614078: 729271 - 738005 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504653.2 XP_003504701.1 1 - 2 415 70

View Full Product Details