Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmx3
Entrez Gene ID
100767995
Gene Name
H6 family homeobox 3
Gene Aliases
I79_018862
Chromosome Location
Chr.Un|NW_003614900: 260006 - 263078 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510537.2 XP_003510585.1 1 - 2 137 53

View Full Product Details