Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mamld1
Entrez Gene ID
100761274
Gene Name
mastermind-like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613974: 1222767 - 1342714 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644086.1 XP_007642276.1 4 - 5 1998 71

View Full Product Details