Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp8
Entrez Gene ID
100759953
Gene Name
matrix metallopeptidase 8 (neutrophil collagenase)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613612: 825472 - 834269 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647375.1 XP_007645565.1 3 - 4 500 79

View Full Product Details