Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stox1
Entrez Gene ID
100771476
Gene Name
storkhead box 1
Gene Aliases
I79_006062
Chromosome Location
Chr.Un|NW_003613759: 1978265 - 1987066 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499910.1 XP_003499958.1 1 - 2 171 72

View Full Product Details