Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndufv3
Entrez Gene ID
100761076
Gene Name
NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614068: 1098113 - 1108280 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504441.2 XP_003504489.1 3 - 4 1337 73
GenBank mRNA JI889204.1 - 3 - 4 1324 73
JP056576.1 - 3 - 4 1324 73

View Full Product Details